Product Details

SNP ID
rs28888397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:213293665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCCTTCAGATGACTCTAATTCT[C/G]CTGTTATTTTGACTATGACATGAGA
Phenotype
MIM: 612173
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100130451 PubMed Links

Gene Details

Gene
LOC100130451
Gene Name
uncharacterized LOC100130451
There are no transcripts associated with this gene.

Gene
SPAG16
Gene Name
sperm associated antigen 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025436.2 Intron NP_001020607.1
NM_024532.4 Intron NP_078808.3
XM_006712746.2 Intron XP_006712809.1
XM_011511814.1 Intron XP_011510116.1
XM_011511815.1 Intron XP_011510117.1
XM_011511816.2 Intron XP_011510118.1
XM_011511817.1 Intron XP_011510119.1
XM_011511818.1 Intron XP_011510120.1
XM_011511819.2 Intron XP_011510121.1
XM_011511820.1 Intron XP_011510122.1
XM_011511821.2 Intron XP_011510123.1
XM_011511822.1 Intron XP_011510124.1
XM_011511823.2 Intron XP_011510125.1
XM_011511824.1 Intron XP_011510126.1
XM_011511827.1 Intron XP_011510129.1
XM_011511828.1 Intron XP_011510130.1
XM_011511829.1 Intron XP_011510131.1
XM_011511831.1 Intron XP_011510133.1
XM_011511832.1 Intron XP_011510134.1
XM_011511835.2 Intron XP_011510137.1
XM_011511837.2 Intron XP_011510139.1
XM_017004896.1 Intron XP_016860385.1
XM_017004897.1 Intron XP_016860386.1
XM_017004898.1 Intron XP_016860387.1
XM_017004899.1 Intron XP_016860388.1
XM_017004900.1 Intron XP_016860389.1
XM_017004901.1 Intron XP_016860390.1
XM_017004902.1 Intron XP_016860391.1

View Full Product Details