Product Details
- SNP ID
-
rs11556885
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:230209959 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTGTCTGAAGGTGTGCTGGACAC[C/T]TCCTGGGGCTCTTCTGGGTCATTTG
- Phenotype
-
MIM: 604457
MIM: 608602
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SP110
PubMed Links
Gene Details
- Gene
- SP110
- Gene Name
- SP110 nuclear body protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001185015.1 |
1041 |
Silent Mutation |
GAA,GAG |
E273E |
NP_001171944.1 |
NM_004509.3 |
1041 |
Silent Mutation |
GAA,GAG |
E267E |
NP_004500.3 |
NM_004510.3 |
1041 |
Silent Mutation |
GAA,GAG |
E267E |
NP_004501.3 |
NM_080424.2 |
1041 |
Silent Mutation |
GAA,GAG |
E267E |
NP_536349.2 |
XM_005246525.3 |
1041 |
Silent Mutation |
GAA,GAG |
E273E |
XP_005246582.1 |
XM_006712487.3 |
1041 |
Silent Mutation |
GAA,GAG |
E273E |
XP_006712550.1 |
XM_006712489.3 |
1041 |
Silent Mutation |
GAA,GAG |
E273E |
XP_006712552.1 |
XM_011511088.2 |
1041 |
Silent Mutation |
GAA,GAG |
E273E |
XP_011509390.1 |
XM_011511089.2 |
1041 |
Silent Mutation |
GAA,GAG |
E267E |
XP_011509391.1 |
XM_011511090.2 |
1041 |
Silent Mutation |
GAA,GAG |
E273E |
XP_011509392.1 |
XM_011511091.2 |
1041 |
Silent Mutation |
GAA,GAG |
E273E |
XP_011509393.1 |
XM_011511092.2 |
1041 |
Silent Mutation |
GAA,GAG |
E64E |
XP_011509394.1 |
XM_017003968.1 |
1041 |
Silent Mutation |
GAA,GAG |
E273E |
XP_016859457.1 |
XM_017003969.1 |
1041 |
Silent Mutation |
GAA,GAG |
E273E |
XP_016859458.1 |
XM_017003970.1 |
1041 |
Silent Mutation |
GAA,GAG |
E267E |
XP_016859459.1 |
- Gene
- SP140
- Gene Name
- SP140 nuclear body protein
View Full Product Details