Product Details

SNP ID
rs3189053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232379666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCGGCAACGAGGTCATCTCCGTG[A/G]TGAATCGGGCCAAGAAAGCAGGTGA
Phenotype
MIM: 171800
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALPP PubMed Links

Gene Details

Gene
ALPP
Gene Name
alkaline phosphatase, placental
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001632.4 732 Missense Mutation ATG,GTG M155V NP_001623.3
Gene
ECEL1P2
Gene Name
endothelin converting enzyme like 1 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details