Product Details

SNP ID
rs28690162
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151197795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGCCTTTATTATAACCTACAG[A/G]TAGAATATAATTACAAATAATACAT
Phenotype
MIM: 611318
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GPR171 PubMed Links

Gene Details

Gene
GPR171
Gene Name
G protein-coupled receptor 171
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013308.3 2216 Intron NP_037440.3
XM_005247402.3 2216 UTR 3 XP_005247459.1
XM_005247403.3 2216 UTR 3 XP_005247460.1
XM_017006274.1 2216 UTR 3 XP_016861763.1
Gene
MED12L
Gene Name
mediator complex subunit 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053002.5 2216 Intron NP_443728.3
XM_006713487.3 2216 Intron XP_006713550.1
XM_011512390.2 2216 Intron XP_011510692.1
XM_011512394.2 2216 Intron XP_011510696.1
XM_011512399.2 2216 Intron XP_011510701.1
XM_017005676.1 2216 Intron XP_016861165.1
XM_017005677.1 2216 Intron XP_016861166.1
XM_017005678.1 2216 Intron XP_016861167.1
XM_017005679.1 2216 Intron XP_016861168.1
XM_017005680.1 2216 Intron XP_016861169.1
XM_017005681.1 2216 Intron XP_016861170.1

View Full Product Details