Product Details
- SNP ID
-
rs35517186
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:187669184 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTCAAGCCTCATTTCATCCTGCT[C/T]AGCAGCCTGGGACAGATCTTCAGGT
- Phenotype
-
MIM: 182450
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SST
PubMed Links
Gene Details
- Gene
- SST
- Gene Name
- somatostatin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001048.3 |
354 |
Missense Mutation |
AAG,GAG |
K78E |
NP_001039.1 |
View Full Product Details