Product Details

SNP ID
rs28564368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:3076335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCTATTTCCGATCTGGATGTGT[A/C]CCAGATGGCATTTGGTAAGAATATC
Phenotype
MIM: 613004
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HTT PubMed Links

Gene Details

Gene
HTT
Gene Name
huntingtin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002111.8 Intron NP_002102.4
Gene
HTT-AS
Gene Name
HTT antisense RNA (head to head)
There are no transcripts associated with this gene.

View Full Product Details