Product Details

SNP ID
rs17884005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1794217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGGCCAGAGCTAGCGCGGCGAC[C/T]TGTGGTGCGCCCGGAGCCGCAGCTA
Phenotype
MIM: 134934
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGFR3 PubMed Links

Gene Details

Gene
FGFR3
Gene Name
fibroblast growth factor receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000142.4 Intron NP_000133.1
NM_001163213.1 Intron NP_001156685.1
NM_022965.3 Intron NP_075254.1
XM_006713868.1 Intron XP_006713931.1
XM_006713869.1 Intron XP_006713932.1
XM_006713870.1 Intron XP_006713933.1
XM_006713871.1 Intron XP_006713934.1
XM_006713872.1 Intron XP_006713935.1
XM_006713873.1 Intron XP_006713936.1
XM_011513420.1 Intron XP_011511722.1
XM_011513422.1 Intron XP_011511724.1

View Full Product Details