Product Details

SNP ID
rs35624275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2073542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGCAACAGTGGCTTTGTGCCAC[A/G]GGCAGGCTGACCCCGTTGTTGGGGC
Phenotype
MIM: 610647 MIM: 610887
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAT8L PubMed Links

Gene Details

Gene
NAT8L
Gene Name
N-acetyltransferase 8 like
There are no transcripts associated with this gene.

Gene
POLN
Gene Name
DNA polymerase nu
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181808.3 Intron NP_861524.2

View Full Product Details