Product Details

SNP ID
rs28687404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:47452182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCACAACAGTTTCCACTGTTTAA[A/G]TATTATTTTTCTCATTTATTAAACA
Phenotype
MIM: 616656
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COMMD8 PubMed Links

Gene Details

Gene
COMMD8
Gene Name
COMM domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017008330.1 1459 UTR 3 XP_016863819.1

View Full Product Details