Product Details
- SNP ID
-
rs28539155
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:47599700 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTGACATTTGAGGACATGCTGGC[A/G]ATCAAGTGAGGGCCCAGACAACACC
- Phenotype
-
MIM: 605236
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP10D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78860216] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP10D
- Gene Name
- ATPase phospholipid transporting 10D (putative)
There are no transcripts associated with this gene.
- Gene
- CORIN
- Gene Name
- corin, serine peptidase
View Full Product Details