Product Details

SNP ID
rs28363897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:24796203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGTCCTCTTTCTCTGATAACAC[C/T]GTCCTCTTTCTCTGATAACACCGTC
Phenotype
MIM: 185490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC149 PubMed Links

Gene Details

Gene
CCDC149
Gene Name
coiled-coil domain containing 149
There are no transcripts associated with this gene.

Gene
SOD3
Gene Name
superoxide dismutase 3, extracellular
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003102.2 Intron NP_003093.2

View Full Product Details