Product Details

SNP ID
rs34948863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15935684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCAAAGACTCTTCGCTGCTCA[A/G]ACACATAGCTGTGTGGGCCATGGAA
Phenotype
MIM: 607737
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGFBP1 PubMed Links

Gene Details

Gene
FGFBP1
Gene Name
fibroblast growth factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005130.4 1244 UTR 3 NP_005121.1

View Full Product Details