Product Details

SNP ID
rs34630753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1254937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCCTGGGTTTGCACAAGGGCCC[C/T]AAGTGCCATGGACGGGAGACAGCCC
Phenotype
MIM: 610300 MIM: 187270
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC6A18 PubMed Links

Gene Details

Gene
SLC6A18
Gene Name
solute carrier family 6 member 18
There are no transcripts associated with this gene.

Gene
TERT
Gene Name
telomerase reverse transcriptase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193376.1 Intron NP_001180305.1
NM_198253.2 Intron NP_937983.2
XM_017009796.1 Intron XP_016865285.1

View Full Product Details