Product Details

SNP ID
rs28424066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:64171836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTTAATTTTTAGAGGCCTTCTC[C/T]ACCCCGCCCCCCCACCAATAACTAA
Phenotype
MIM: 616015
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF180 PubMed Links

Gene Details

Gene
RNF180
Gene Name
ring finger protein 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113561.2 Intron NP_001107033.1
NM_001323291.1 Intron NP_001310220.1
NM_001323292.1 Intron NP_001310221.1
NM_178532.4 Intron NP_848627.1
XM_017009383.1 Intron XP_016864872.1
XM_017009384.1 Intron XP_016864873.1
XM_017009385.1 Intron XP_016864874.1
XM_017009386.1 Intron XP_016864875.1
XM_017009387.1 Intron XP_016864876.1
XM_017009388.1 Intron XP_016864877.1
XM_017009389.1 Intron XP_016864878.1

View Full Product Details