Product Details

SNP ID
rs28608884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69376288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGTGCTGTAGCTTCAGTTTTTC[A/C]ATCCTTGCCTGAATCACTTATTTTA
Phenotype
MIM: 603139 MIM: 600822
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AK6 PubMed Links

Gene Details

Gene
AK6
Gene Name
adenylate kinase 6
There are no transcripts associated with this gene.

Gene
RAD17
Gene Name
RAD17 checkpoint clamp loader component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278622.1 Intron NP_001265551.1
NM_002873.1 Intron NP_002864.1
NM_133338.2 Intron NP_579916.1
NM_133339.2 Intron NP_579917.1
NM_133340.2 Intron NP_579918.1
NM_133341.2 Intron NP_579919.1
NM_133342.2 Intron NP_579920.1
NM_133343.1 Intron NP_579921.1
NM_133344.2 Intron NP_579922.1
XM_017009679.1 Intron XP_016865168.1
XM_017009680.1 Intron XP_016865169.1
XM_017009681.1 Intron XP_016865170.1
Gene
TAF9
Gene Name
TATA-box binding protein associated factor 9
There are no transcripts associated with this gene.

View Full Product Details