Product Details

SNP ID
rs28447466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:71010851 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCCTGCTTGAATAAATCCTGGAG[C/T]GGTCCAATTTTGAATAGGGAAAGAT
Phenotype
MIM: 600355
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAIP PubMed Links

Gene Details

Gene
NAIP
Gene Name
NLR family apoptosis inhibitory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004536.2 Intron NP_004527.2
NM_022892.1 Intron NP_075043.1
XM_011543413.2 Intron XP_011541715.1
XM_017009490.1 Intron XP_016864979.1

View Full Product Details