Product Details

SNP ID
rs28382594
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28526877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTATGGGTAACAGTAACTACCT[C/T]ATGGGTGGGTGAATAACAATTATTA
Phenotype
MIM: 603435
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GPX5 PubMed Links

Gene Details

Gene
GPX5
Gene Name
glutathione peroxidase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001509.2 Intron NP_001500.1
NM_003996.3 Intron NP_003987.2

View Full Product Details