Product Details

SNP ID
rs41541412
Assay Type
Functionally Tested
NCBI dbSNP Submissions
4
Location
Chr.6:32641472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGCAAATTTGGAGGTTTTGACC[C/G]GCAGGGTGCACTGAGAAACATGGCT
Phenotype
MIM: 146880
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
HLA-DQA1 PubMed Links

Gene Details

Gene
HLA-DQA1
Gene Name
major histocompatibility complex, class II, DQ alpha 1
There are no transcripts associated with this gene.

View Full Product Details