Product Details

SNP ID
rs28453017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42053092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATATTTATTTATTCTTATTAATT[A/G]GAGATGGGGTCTTGCTGTGTTGCCC
Phenotype
MIM: 123834 MIM: 609514
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCND3 PubMed Links

Gene Details

Gene
CCND3
Gene Name
cyclin D3
There are no transcripts associated with this gene.

Gene
TAF8
Gene Name
TATA-box binding protein associated factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138572.2 Intron NP_612639.2
XM_005248849.2 Intron XP_005248906.1
XM_011514296.1 Intron XP_011512598.1
XM_017010241.1 Intron XP_016865730.1
XM_017010242.1 Intron XP_016865731.1
XM_017010243.1 Intron XP_016865732.1
XM_017010244.1 Intron XP_016865733.1
XM_017010245.1 Intron XP_016865734.1

View Full Product Details