Product Details

SNP ID
rs28370564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:47861382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTAGTATTTATTTCTTTTGGGT[A/G]AAATTATGAGTTTGGGAACCTATTT
Phenotype
MIM: 616908
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTCHD4 PubMed Links

Gene Details

Gene
PTCHD4
Gene Name
patched domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013732.3 20506 Intron NP_001013754.3
NM_207499.2 20506 Intron NP_997382.2
XM_011514639.2 20506 UTR 3 XP_011512941.1
XM_017010891.1 20506 UTR 3 XP_016866380.1
XM_017010892.1 20506 UTR 3 XP_016866381.1
XM_017010893.1 20506 UTR 3 XP_016866382.1
XM_017010894.1 20506 UTR 3 XP_016866383.1
XM_017010895.1 20506 UTR 3 XP_016866384.1
XM_017010896.1 20506 Intron XP_016866385.1

View Full Product Details