Product Details

SNP ID
rs11545322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:157540765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTCACCTCCTCAGCCACGGCTGT[A/C]AGCGCGAACTCAAACTGCTCCTGCA
Phenotype
MIM: 601698
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTPRN2 PubMed Links

Gene Details

Gene
PTPRN2
Gene Name
protein tyrosine phosphatase, receptor type N2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308267.1 3130 Silent Mutation CTG,CTT L961L NP_001295196.1
NM_001308268.1 3130 Silent Mutation CTG,CTT L1022L NP_001295197.1
NM_002847.4 3130 Silent Mutation CTG,CTT L999L NP_002838.2
NM_130842.3 3130 Silent Mutation CTG,CTT L982L NP_570857.2
NM_130843.3 3130 Silent Mutation CTG,CTT L970L NP_570858.2
XM_011516446.1 3130 Intron XP_011514748.1
XM_011516447.2 3130 Intron XP_011514749.1
XM_011516448.2 3130 Intron XP_011514750.1
XM_011516449.2 3130 Intron XP_011514751.1
XM_017012475.1 3130 Silent Mutation CTG,CTT L933L XP_016867964.1
XM_017012476.1 3130 Intron XP_016867965.1

View Full Product Details