Product Details

SNP ID
rs28664975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44063656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGCGGAATGGAGCTGGAGGAGG[C/G]ACAGGGAGGCCGAGCCTGGTCGAAA
Phenotype
MIM: 610106 MIM: 612931 MIM: 606344
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DBNL PubMed Links

Gene Details

Gene
DBNL
Gene Name
drebrin like
There are no transcripts associated with this gene.

Gene
MIR6838
Gene Name
microRNA 6838
There are no transcripts associated with this gene.

Gene
PGAM2
Gene Name
phosphoglycerate mutase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000290.3 Intron NP_000281.2
Gene
POLM
Gene Name
DNA polymerase mu
There are no transcripts associated with this gene.

View Full Product Details