Product Details

SNP ID
rs35118895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6375095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCGCTCGTAGAGCAAGCGCTCTT[A/G]GAGATTTTGACCATTTTTCCAAAAA
Phenotype
MIM: 602048
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAC1 PubMed Links

Gene Details

Gene
RAC1
Gene Name
ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006908.4 Intron NP_008839.2
NM_018890.3 Intron NP_061485.1

View Full Product Details