Product Details
- SNP ID
-
rs28446721
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:6595572 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGATACCCATTTGCCTGAGGTGCA[A/G]TGGTCCAAGTGGCAGGCAGCCCAGA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C7orf26
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114855642] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C7orf26
- Gene Name
- chromosome 7 open reading frame 26
- Gene
- ZDHHC4
- Gene Name
- zinc finger DHHC-type containing 4
There are no transcripts associated with this gene.
View Full Product Details