Product Details

SNP ID
rs28600681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93100709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTTGATAAGCTAATTAGCTATA[A/G]AGACAAATTTCATTTTTGCTTAACT
Phenotype
MIM: 610456
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAMD9 PubMed Links

Gene Details

Gene
SAMD9
Gene Name
sterile alpha motif domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193307.1 5558 UTR 3 NP_001180236.1
NM_017654.3 5558 UTR 3 NP_060124.2

View Full Product Details