Product Details

SNP ID
rs28595886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99146999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCGTAAGTCAAACCGTCATAAAC[G/T]GGGACCATTTGAAATTGCTAGATAT
Phenotype
MIM: 614107 MIM: 605568
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KPNA7 PubMed Links

Gene Details

Gene
KPNA7
Gene Name
karyopherin subunit alpha 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011516215.2 Intron XP_011514517.1
XM_017012208.1 Intron XP_016867697.1
XM_017012209.1 Intron XP_016867698.1
XM_017012210.1 Intron XP_016867699.1
XM_017012211.1 Intron XP_016867700.1
XM_017012212.1 Intron XP_016867701.1
XM_017012213.1 Intron XP_016867702.1
XM_017012214.1 Intron XP_016867703.1
XM_017012215.1 Intron XP_016867704.1
XM_017012216.1 Intron XP_016867705.1
Gene
SMURF1
Gene Name
SMAD specific E3 ubiquitin protein ligase 1
There are no transcripts associated with this gene.

View Full Product Details