Product Details
- SNP ID
-
rs28373136
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:101206688 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAAAGTCACTGGGAAATGGGCAGG[A/G]AGGGGAGCTGGGGGATGCACGCAGG
- Phenotype
-
MIM: 610184
MIM: 603066
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MOGAT3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80179986] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MOGAT3
- Gene Name
- monoacylglycerol O-acyltransferase 3
There are no transcripts associated with this gene.
- Gene
- PLOD3
- Gene Name
- procollagen-lysine,2-oxoglutarate 5-dioxygenase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001084.4 |
|
Intron |
|
|
NP_001075.1 |
View Full Product Details