Product Details

SNP ID
rs28657394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:105462097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGATAGAAAAGGAAAAGAAAA[A/G]GAAAAGGAAAAGAAAAGAAGATAAA
Phenotype
MIM: 616261
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PUS7 PubMed Links

Gene Details

Gene
PUS7
Gene Name
pseudouridylate synthase 7 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318163.1 Intron NP_001305092.1
NM_001318164.1 Intron NP_001305093.1
NM_019042.4 Intron NP_061915.2
XM_005250462.3 Intron XP_005250519.1
XM_011516336.2 Intron XP_011514638.1
XM_017012367.1 Intron XP_016867856.1
XM_017012368.1 Intron XP_016867857.1

View Full Product Details