Product Details

SNP ID
rs1131358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:106090529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTTTCAACTCCTTTATTAAAT[G/T]GGTTGCAAAAAAGATATACATTCTT
Phenotype
MIM: 616665
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SYPL1 PubMed Links

Gene Details

Gene
SYPL1
Gene Name
synaptophysin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006754.3 2174 UTR 3 NP_006745.1
NM_182715.2 2174 UTR 3 NP_874384.1

View Full Product Details