Product Details

SNP ID
rs1803093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:134445282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGTTCCTGTTGTAGCTGAGTAA[A/G]GTGGTCATATCCTGGCTGCTCAGTT
Phenotype
MIM: 103880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKR1B1 PubMed Links

Gene Details

Gene
AKR1B1
Gene Name
aldo-keto reductase family 1 member B
There are no transcripts associated with this gene.

View Full Product Details