Product Details
- SNP ID
-
rs36031925
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:37965413 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGAAGACGGCGGAAGGCGCTGC[A/G]AAAGTCCGGGCTGCGGCAGTAGATG
- Phenotype
-
MIM: 109691
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADRB3
PubMed Links
Gene Details
- Gene
- ADRB3
- Gene Name
- adrenoceptor beta 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000025.2 |
1254 |
Missense Mutation |
|
|
NP_000016.1 |
View Full Product Details