Product Details
- SNP ID
-
hCV59146926
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:30678395 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTTTTGAGACAGGGTCTTGCTC[C/T]GTTGTCCAAGATTGAGTGCAGCAGC
- Phenotype
-
MIM: 138300
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GSR
PubMed Links
Gene Details
- Gene
- GSR
- Gene Name
- glutathione-disulfide reductase
There are no transcripts associated with this gene.
View Full Product Details