Product Details

SNP ID
rs28404611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39591302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCTTTCATGAAAGATTTCTCT[A/G]TAGTATGCAATGCTGTTTGATAGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM18 PubMed Links

Gene Details

Gene
ADAM18
Gene Name
ADAM metallopeptidase domain 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190956.1 Intron NP_001177885.1
NM_001320313.1 Intron NP_001307242.1
NM_014237.2 Intron NP_055052.1

View Full Product Details