Product Details

SNP ID
rs1136495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39744055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTCCATCCATCACAGATAATTA[C/G]TCTAGAAGACTGAATAAATGAAAAA
Phenotype
MIM: 601533
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADAM2 PubMed Links

Gene Details

Gene
ADAM2
Gene Name
ADAM metallopeptidase domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278113.1 2200 UTR 3 NP_001265042.1
NM_001278114.1 2200 UTR 3 NP_001265043.1
NM_001464.4 2200 UTR 3 NP_001455.3
XM_005273468.1 2200 UTR 3 XP_005273525.1
XM_011544479.1 2200 UTR 3 XP_011542781.1
XM_011544480.1 2200 UTR 3 XP_011542782.1

View Full Product Details