Product Details

SNP ID
rs28357249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:117803662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAATTCCCAATTCTGTCTCTAAC[C/T]TACTGGGCAACTTGTGGCAATATGC
Phenotype
MIM: 608177
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXT1 PubMed Links

Gene Details

Gene
EXT1
Gene Name
exostosin glycosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000127.2 Intron NP_000118.2

View Full Product Details