Product Details

SNP ID
rs17432076
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:118136749 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTGTAGATGTGGAAACCAGGGTA[A/G]GGGAGTCACTTTAATGTCTGCTGTA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SAMD12 PubMed Links

Gene Details

Gene
SAMD12
Gene Name
sterile alpha motif domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101676.1 Intron NP_001095146.1
NM_207506.2 Intron NP_997389.2
XM_011517031.2 Intron XP_011515333.1
XM_011517034.2 Intron XP_011515336.1
XM_011517035.2 Intron XP_011515337.1
XM_017013409.1 Intron XP_016868898.1
XM_017013410.1 Intron XP_016868899.1
XM_017013411.1 Intron XP_016868900.1
XM_017013412.1 Intron XP_016868901.1

View Full Product Details