Product Details

SNP ID
rs28585677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:91114800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGACAAAATTGCCTAACGATGC[A/G]TTTCTTAGAATGTGTCCCCATAGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC69 PubMed Links

Gene Details

Gene
LRRC69
Gene Name
leucine rich repeat containing 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129890.1 Intron NP_001123362.1

View Full Product Details