Product Details

SNP ID
rs28475202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:140661412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGGTGTGTGGTTGTTTGCATG[C/T]GTGTAGGGGATTGGGAACAGACATG
Phenotype
MIM: 600758
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTK2 PubMed Links

Gene Details

Gene
PTK2
Gene Name
protein tyrosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199649.1 Intron NP_001186578.1
NM_001316342.1 Intron NP_001303271.1
NM_005607.4 Intron NP_005598.3
NM_153831.3 Intron NP_722560.1
XM_005251003.2 Intron XP_005251060.1
XM_005251012.2 Intron XP_005251069.1
XM_005251013.2 Intron XP_005251070.1
XM_006716608.2 Intron XP_006716671.1
XM_006716609.2 Intron XP_006716672.1
XM_006716610.2 Intron XP_006716673.1
XM_011517194.2 Intron XP_011515496.1
XM_017013651.1 Intron XP_016869140.1
XM_017013652.1 Intron XP_016869141.1
XM_017013653.1 Intron XP_016869142.1
XM_017013654.1 Intron XP_016869143.1
XM_017013655.1 Intron XP_016869144.1
XM_017013656.1 Intron XP_016869145.1
XM_017013657.1 Intron XP_016869146.1
XM_017013658.1 Intron XP_016869147.1
XM_017013659.1 Intron XP_016869148.1
XM_017013660.1 Intron XP_016869149.1
XM_017013661.1 Intron XP_016869150.1
XM_017013662.1 Intron XP_016869151.1
XM_017013663.1 Intron XP_016869152.1
XM_017013664.1 Intron XP_016869153.1
XM_017013665.1 Intron XP_016869154.1
XM_017013666.1 Intron XP_016869155.1
XM_017013667.1 Intron XP_016869156.1
XM_017013668.1 Intron XP_016869157.1
XM_017013669.1 Intron XP_016869158.1
XM_017013670.1 Intron XP_016869159.1
XM_017013671.1 Intron XP_016869160.1
XM_017013672.1 Intron XP_016869161.1
XM_017013673.1 Intron XP_016869162.1
XM_017013674.1 Intron XP_016869163.1
XM_017013675.1 Intron XP_016869164.1
XM_017013676.1 Intron XP_016869165.1
XM_017013677.1 Intron XP_016869166.1
XM_017013678.1 Intron XP_016869167.1
XM_017013679.1 Intron XP_016869168.1
XM_017013680.1 Intron XP_016869169.1
XM_017013681.1 Intron XP_016869170.1
XM_017013682.1 Intron XP_016869171.1
XM_017013683.1 Intron XP_016869172.1
XM_017013684.1 Intron XP_016869173.1
XM_017013685.1 Intron XP_016869174.1
XM_017013686.1 Intron XP_016869175.1
XM_017013687.1 Intron XP_016869176.1
XM_017013688.1 Intron XP_016869177.1
XM_017013689.1 Intron XP_016869178.1
XM_017013690.1 Intron XP_016869179.1

View Full Product Details