Product Details

SNP ID
rs28639943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86515328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTATAATGGATAATCAAATAGA[A/G]GGAAGGGCGGGATTGAATATTGTGA
Phenotype
MIM: 604207 MIM: 611871
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPNE3 PubMed Links

Gene Details

Gene
CPNE3
Gene Name
copine 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003909.4 Intron NP_003900.1
XM_005251093.4 Intron XP_005251150.1
XM_017013945.1 Intron XP_016869434.1
Gene
RMDN1
Gene Name
regulator of microtubule dynamics 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286707.1 Intron NP_001273636.1
NM_001286719.1 Intron NP_001273648.1
NM_001317807.1 Intron NP_001304736.1
NM_016033.2 Intron NP_057117.2
XM_005250944.3 Intron XP_005251001.1
XM_005250947.3 Intron XP_005251004.1
XM_005250948.3 Intron XP_005251005.1
XM_006716573.3 Intron XP_006716636.1
XM_011517090.2 Intron XP_011515392.1
XM_011517091.2 Intron XP_011515393.1
XM_011517092.2 Intron XP_011515394.1
XM_011517093.2 Intron XP_011515395.1
XM_011517094.2 Intron XP_011515396.1
XM_011517095.2 Intron XP_011515397.1
XM_011517096.2 Intron XP_011515398.1
XM_011517097.2 Intron XP_011515399.1
XM_011517098.2 Intron XP_011515400.1
XM_017013518.1 Intron XP_016869007.1
XM_017013519.1 Intron XP_016869008.1
XM_017013520.1 Intron XP_016869009.1
XM_017013521.1 Intron XP_016869010.1
XM_017013522.1 Intron XP_016869011.1
XM_017013523.1 Intron XP_016869012.1
XM_017013524.1 Intron XP_016869013.1
XM_017013525.1 Intron XP_016869014.1
XM_017013526.1 Intron XP_016869015.1
XM_017013527.1 Intron XP_016869016.1
XM_017013528.1 Intron XP_016869017.1

View Full Product Details