Product Details

SNP ID
rs28399587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94881362 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTAAAGGAAAAACATTGCTATGGT[A/G]TAGACTGTGGTTGGCTTCTATCCAG
Phenotype
MIM: 603775 MIM: 611351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNE2 PubMed Links

Gene Details

Gene
CCNE2
Gene Name
cyclin E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057749.2 1914 UTR 3 NP_477097.1
XM_011517366.2 1914 Intron XP_011515668.1
XM_017013957.1 1914 UTR 3 XP_016869446.1
XM_017013958.1 1914 UTR 3 XP_016869447.1
XM_017013959.1 1914 UTR 3 XP_016869448.1
Gene
INTS8
Gene Name
integrator complex subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017864.3 1914 Intron NP_060334.2
XM_017013616.1 1914 Intron XP_016869105.1
XM_017013617.1 1914 Intron XP_016869106.1
XM_017013618.1 1914 UTR 3 XP_016869107.1
XM_017013619.1 1914 UTR 3 XP_016869108.1

View Full Product Details