Product Details

SNP ID
rs28399553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94894566 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTCCTCCACCCTCTCCCCCGTAC[A/G]CCGGGAGTGGGGCTTGGGCAGCGCC
Phenotype
MIM: 603775 MIM: 611351 MIM: 612392
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNE2 PubMed Links

Gene Details

Gene
CCNE2
Gene Name
cyclin E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057749.2 85 Intron NP_477097.1
XM_011517366.2 85 UTR 5 XP_011515668.1
XM_017013957.1 85 UTR 5 XP_016869446.1
XM_017013958.1 85 Intron XP_016869447.1
XM_017013959.1 85 Intron XP_016869448.1
Gene
INTS8
Gene Name
integrator complex subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017864.3 85 Intron NP_060334.2
XM_017013616.1 85 UTR 3 XP_016869105.1
XM_017013617.1 85 Intron XP_016869106.1
XM_017013618.1 85 Intron XP_016869107.1
XM_017013619.1 85 Intron XP_016869108.1
Gene
NDUFAF6
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152416.3 85 Intron NP_689629.2
XM_005250789.2 85 Intron XP_005250846.1
XM_005250790.2 85 Intron XP_005250847.1
XM_005250791.2 85 Intron XP_005250848.1
XM_005250792.2 85 Intron XP_005250849.1
XM_005250793.2 85 Intron XP_005250850.1
XM_011516833.2 85 Intron XP_011515135.1
XM_011516834.2 85 Intron XP_011515136.1
XM_011516835.2 85 Intron XP_011515137.1
XM_011516836.2 85 Intron XP_011515138.1
XM_011516837.2 85 Intron XP_011515139.1
XM_011516838.2 85 Intron XP_011515140.1
XM_011516839.2 85 Intron XP_011515141.1
XM_011516840.2 85 Intron XP_011515142.1
XM_011516841.2 85 Intron XP_011515143.1
XM_011516842.2 85 Intron XP_011515144.1
XM_017013027.1 85 Intron XP_016868516.1
XM_017013028.1 85 Intron XP_016868517.1
XM_017013029.1 85 Intron XP_016868518.1
XM_017013030.1 85 Intron XP_016868519.1
XM_017013031.1 85 Intron XP_016868520.1
XM_017013032.1 85 Intron XP_016868521.1
XM_017013033.1 85 Intron XP_016868522.1
XM_017013034.1 85 Intron XP_016868523.1
XM_017013035.1 85 Intron XP_016868524.1
XM_017013036.1 85 Intron XP_016868525.1
XM_017013037.1 85 Intron XP_016868526.1
XM_017013038.1 85 Intron XP_016868527.1
XM_017013039.1 85 Intron XP_016868528.1

View Full Product Details