Product Details
- SNP ID
-
rs34066384
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21441666 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAACTAATGATTTCTCTCAGCAGC[A/G]GGTAAACTGAGACAGGTGAGGAAAT
- Phenotype
-
MIM: 147660
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IFNA1
PubMed Links
Gene Details
- Gene
- IFNA1
- Gene Name
- interferon alpha 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024013.2 |
|
Intron |
|
|
NP_076918.1 |
View Full Product Details