Product Details

SNP ID
rs34433332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:2622077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCAACTCCTTCCCCTCCTTCTCC[C/G]CCTTTCCCCTCCCCGCCCCCACCTT
Phenotype
MIM: 192977
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
VLDLR PubMed Links

Gene Details

Gene
VLDLR
Gene Name
very low density lipoprotein receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018056.2 304 UTR 5 NP_001018066.1
NM_001322225.1 304 UTR 5 NP_001309154.1
NM_001322226.1 304 UTR 5 NP_001309155.1
NM_003383.4 304 UTR 5 NP_003374.3
Gene
VLDLR-AS1
Gene Name
VLDLR antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details