Product Details

SNP ID
rs28547054
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34614295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTGCCCTAACAGCAGTCCAAAG[A/G]CGGATGGAAAAGGGAGTGGCCATAC
Phenotype
MIM: 607387
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARID3C PubMed Links

Gene Details

Gene
ARID3C
Gene Name
AT-rich interaction domain 3C
There are no transcripts associated with this gene.

Gene
DCTN3
Gene Name
dynactin subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281425.1 Intron NP_001268354.1
NM_001281426.1 Intron NP_001268355.1
NM_001281427.1 Intron NP_001268356.1
NM_007234.4 Intron NP_009165.1
NM_024348.3 Intron NP_077324.1
Gene
RPP25L
Gene Name
ribonuclease P/MRP subunit p25 like
There are no transcripts associated with this gene.

View Full Product Details