Product Details

SNP ID
rs28361184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:108942629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAAAAGCTTTACAATCAGTTTC[A/G]TGATCAGAAAATAGAGCAAAATTTC
Phenotype
MIM: 604785 MIM: 603722
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTNNAL1 PubMed Links

Gene Details

Gene
CTNNAL1
Gene Name
catenin alpha like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286974.1 2534 UTR 3 NP_001273903.1
NM_003798.3 2534 UTR 3 NP_003789.1
XM_005252291.3 2534 UTR 3 XP_005252348.1
XM_017015250.1 2534 UTR 3 XP_016870739.1
XM_017015251.1 2534 UTR 3 XP_016870740.1
Gene
FAM206A
Gene Name
family with sequence similarity 206 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017832.3 2534 Intron NP_060302.1
XM_011518816.2 2534 Intron XP_011517118.1
Gene
IKBKAP
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
There are no transcripts associated with this gene.

View Full Product Details