Product Details
- SNP ID
-
rs34786778
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:125237694 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATTAAAAAAAAAAAAAAAAGAATG[T/C]TAAATATCTAATATCAACATTATTC
- Phenotype
-
MIM: 138120
MIM: 605962
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HSPA5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35644223] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSPA5
- Gene Name
- heat shock protein family A (Hsp70) member 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005347.4 |
|
Intron |
|
|
NP_005338.1 |
- Gene
- RABEPK
- Gene Name
- Rab9 effector protein with kelch motifs
There are no transcripts associated with this gene.
View Full Product Details