Product Details

SNP ID
rs17508293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129038278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGCGAGACTGACAGGCTCCTCA[C/G]TCCTTCCCACTCACTGGGGAGAAGC
Phenotype
MIM: 616774 MIM: 609288
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIGA2 PubMed Links

Gene Details

Gene
MIGA2
Gene Name
mitoguardin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005252282.1 Intron XP_005252339.1
XM_011519117.1 Intron XP_011517419.1
XM_011519118.1 Intron XP_011517420.1
Gene
SH3GLB2
Gene Name
SH3 domain containing GRB2 like endophilin B2
There are no transcripts associated with this gene.

View Full Product Details