Product Details

SNP ID
rs35854553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:88176505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTCCACCTACAAGAAGGTAAA[A/T]TTTTTTGAGGCCAGTAGGCATTGTC
Phenotype
MIM: 606281
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR3166 PubMed Links

Gene Details

Gene
MIR3166
Gene Name
microRNA 3166
There are no transcripts associated with this gene.

Gene
RAB38
Gene Name
RAB38, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details