Product Details

SNP ID
hCV59698662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94417288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCACCCAGCCCATTGAGATAC[C/T]TTTTTACTCAGTTTTTTTCTAAGAA
Phenotype
MIM: 600814
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRE11A PubMed Links

Gene Details

Gene
MRE11A
Gene Name
MRE11 homolog A, double strand break repair nuclease
There are no transcripts associated with this gene.

View Full Product Details