Product Details

SNP ID
rs28757318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2618604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCATACTGTTTGATCAATAGTTT[G/T]GTAAAATAAACCAGAAAGTATGAAG
Phenotype
MIM: 607542 MIM: 604115
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNQ1 PubMed Links

Gene Details

Gene
KCNQ1
Gene Name
potassium voltage-gated channel subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000218.2 Intron NP_000209.2
NM_181798.1 Intron NP_861463.1
Gene
KCNQ1OT1
Gene Name
KCNQ1 opposite strand/antisense transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details